4'-Methylacetanilide, 98+%
4'-Methylacetanilide, 98+%
4'-Methylacetanilide, 98+%
Thermo Scientific Chemicals

4'-Methylacetanilide, 98+%

CAS: 103-89-9 | C9H11NO | 149.193 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A13721.14
also known as A13721-14
Price (USD)
25.90
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
25.90
Each
Chemical Identifiers
CAS103-89-9
IUPAC NameN-(4-methylphenyl)acetamide
Molecular FormulaC9H11NO
InChI KeyYICAMJWHIUMFDI-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown or grey
FormCrystals or powder or crystalline powder
Melting Point (clear melt)145.0-151.0?C
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only