ExpressLink™ T4 DNA Ligase - FAQs
ExpressLink™ T4 DNA Ligase - FAQs
View additional product information for ExpressLink™ T4 DNA Ligase - FAQs (A13726)
30 product FAQs found
What kind of controls should I have for restriction cloning?
You can have all of the below controls or select the one you consider the most appropriate to the problem you are facing:
1– Transform the E. coli with circular plasmid to assess the competency of the cells (how well they are taking up DNA).
2– Transform and plate the dephosphorylated vector. It will help you assess how well the dephosphorylation worked and what proportion of colonies in your ligation transformation plate could be false positives (re-ligated vector or background).
3– Use T4 DNA igase to re-ligate your cut vector, or lambda DNA/Hind III marker. It will help you assess whether the ligase itself is working properly.
What are common inhibitors of the T4 DNA ligase?
dATP is a competitive inhibitor. Phosphate will reduce ligation efficiency. Detergents in your ligation buffer will likely not affect activity. High levels (0.2M) Na2+, K+, Cs+, Li+, and NH4+ inhibit the enzyme almost completely. Polyamines, spermine, and spermidine also serve as inhibitors.
Do both my insert and vector have to be phosphorylated for successful ligation?
At least one molecule in a ligase reaction (i.e., insert or vector) must be phosphorylated. Ligation reactions are dependent on the presence of a 5' phosphate on the DNA molecules. The ligation of a dephosphorylated vector with an insert generated from a restriction enzyme digest (phosphorylated) is most routinely performed. Although only one strand of the DNA ligates at a junction point, the molecule can form a stable circle, providing that the insert is large enough for hybridization to maintain the molecule in a circular form.
Which T4 DNA Ligase protocol do you recommend when ligating an insert containing one cohesive (sticky) end and one blunt end?
For cloning an insert with one cohesive end and one blunt end, use the conditions for blunt ends.  The sticky end may ligate quickly, but the blunt end ligation will still be inefficient. You should use the more stringent protocol to optimize the blunt end ligation. This usually means using more enzyme (5 U), a lower reaction temperature (14C) and a longer incubation time (16-24 hours). 
Why do I sometimes get low cDNA yield when using SuperScript Reverse Transcriptase?
Low cDNA yield can result due to several different reasons. Please see a few listed below:
(1) Poor quality mRNA: visualize total RNA on a denaturing gel to verify that the 28S and 18S bands are sharp. OD 260:280 ratio should be 1.7.
(2) Template degraded by RNase contamination: maintain aseptic conditions.
(3) Inhibitors of SuperScript II RT may be present: remove inhibitors by ethanol precipitation of the RNA preparation before the first-strand reaction. Include a 70% (v/v) ethanol wash of the RNA pellet. Test for the presence of inhibitors by mixing 1 µg control RNA and comparing yields of first-strand cDNA.
(4) RNA preparation may have coprecipitated with polysaccharides: precipitate RNA with lithium chloride to purify RNA.
(5) mRNA concentrations were overestimated: quantitate the mRNA concentrations by measuring the A260 if possible.
(6) If using 32P-isotope, it may be too old: use isotope less than 2 weeks old.
(7) Not enough enzyme was used: use 200 U SuperScript II RT/µg RNA.
(8) SuperScript II RT activity was decreased by incorrect reaction temperature: perform the first-strand reaction at a temperature between 37 degrees C and 50 degrees C.
(9) DTT was not added to first-strand reaction.
(10) TCA precipitations were performed incorrectly: adequately dry GF/C filters before immersion into scintillant.
(11) SuperScript II RT was improperly stored: store at -20 degrees C. Do not store the enzyme at -70 degrees C.
(12) The reaction volume was too large: the reaction should be done in volumes less than or equal to 50 µL.
Do both my insert and vector need to be phosphorylated for ligation?
At least one molecule in a ligase reaction (i.e., insert or vector) must be phosphorylated. Ligation reactions are dependent on the presence of a 5' phosphate on the DNA molecules. The ligation of a dephosphorylated vector with an insert generated from a restriction enzyme digest (phosphorylated) is most routinely performed. Although only one strand of the DNA ligates at a junction point, the molecule can form a stable circle, providing that the insert is large enough for hybridization to maintain the molecule in a circular form.
What concentration of insert and vector do you recommend for most ligation reactions?
Recommendations would vary depending on the size of the vector and insert or the nature of the insert, but for most plasmid cloning or subcloning reactions, a vector concentration of 1-10 ng/µl is recommended. Inserts should generally be 2- to 3-fold excess in molar concentration relative to the vector.
How can I test for inhibitors in  my ligation reactions?
To test for the presence of ligation inhibitors, perform a ligation reaction in which some of the vector or insert DNA is included along with some marker DNA such as lambda DNA/Hind III Fragments. If ligation of the DNA marker fragments occurs alone but is not observed when other DNA is added, then a diffusible inhibitor is present in the vector or insert DNA.
To purify and remove inhibitors, extract the DNA with buffer-saturated phenol, then extract with chloroform:isoamyl alcohol, and precipitate with ammonium acetate and ethanol. Be sure that the DNA is free of phenol and that the phosphate concentration is less than 25 mM and the NaCl concentration is less than 50 mM. Also, be sure that the DNA is free of contaminating DNA that might compete for ligation to the insert or vector (e.g., linker fragments, DNA fragments from which the insert was completely purified).
If restriction endonucleases are present, causing redigestion of ligated products, your ligation will also be inhibited. After digestion of the vector and insert DNA, remove restriction endonucleases by extraction with buffer-saturated phenol, extraction with chloroform:isoamyl alcohol, and ethanol precipitation.
How can I determine if my ligase is still active?
Ligation reagents may be tested by performing a ligation reaction with a molecular size marker such as the 1 Kb DNA Ladder or lambda DNA/Hind III Fragments. Compare the ligation reaction products to unligatedDNA on an agarose gel. The ligation reaction should contain a high molecular weight smear and few low-molecular weight bands. If the marker ligation does not work, use fresh ligase.
Another reason for low activity could be degradation of the ATP in the reaction buffer; use 5X ligase buffer that is less than 24 months old. The buffer should be stored at -20 degrees C.
If ligation/transformation reactions did not yield any colonies, or if a high number of background colonies are observed, what control reactions should be used to determine the problem?
Several ligation controls may be necessary to identify the source of ligation problems.
Recommendations for problems with no colonies after transformation:
1. Test the transformation efficiency of the competent cells using a supercoiled vector, or the control DNA provided with Invitrogen competent cells. Perform a transformation reaction and plate the number of cells that is expected to generate 50-100 colonies per plate, based on the anticipated transformation efficiency of the competent cells. The expected number of colonies should be seen, indicating that the competent cells are transforming with high efficiency. The control DNA provided with Invitrogen competent cells is supercoiled monomer; vector DNA preparations that contain other forms will not transform as efficiently. Transformation efficiencies will be up to 10-fold lower for ligated vectors than for intact control DNA.
2. Restriction endonuclease-digested, re-ligated vector. Set up a ligation reaction using the same amount of vector DNA that is used in the experimental ligations and use it to transform competent cells. Re-ligation of vectors with cohesive ends should result in less than or equal to 50% of the number of colonies obtained with supercoiled vector DNA, indicating that the components of the ligation reaction are working; re-ligation of vectors with blunt ends should yield 10% to 20% of the number of colonies obtained with supercoiled vector DNA. This is an appropriate control only with vectors that have been digested with a single restriction endonuclease; double-digested vectors may not re-ligate because the ends are incompatible and the small DNA fragment that is released from between the two sites is sometimes lost during ethanol precipitation of the DNA.
For observation of a high number of background colonies:
1. Restriction endonuclease-digested vector. Perform a transformation with an amount of restriction enzyme-digested vector DNA equivalent to that contained in the fraction of the ligation reaction being used for the experimental transformations. Few or no colonies should be seen, indicating complete restriction endonuclease digestion of the vector. The presence of colonies indicates incomplete digestion of the vector that will cause a background of colonies containing non-recombinant vector in the experimental transformations.
2. Restriction endonuclease-digested, dephosphorylated, re-ligated vector. Set up a ligation reaction using the same amount of vector DNA that is used in the experimental ligations and use it to transform competent cells. Few or no colonies should be observed, indicating complete dephosphorylation of the vector - a dephosphorylated vector should not be re-ligated by T4 DNA ligase.
3. No DNA transformation control. Perform a mock transformation of competent cells, to which no DNA is added. No colonies should be seen, indicating that the selection antibiotic on the agar plates is potent and that the competent cells are pure.
How can I check that my ligation reaction worked?
Ligation reactions are best analyzed by actual transformation of bacteria, since not all of the high molecular weight forms created in a reaction (seen in gel analysis) will transform cells efficiently.
Why is it necessary to dilute ligated DNA products before adding them to competent bacterial cells?
Components of the ligation reaction (enzymes, salts) can interfere with transformation, and may reduce the number of recombinant colonies or plaques. We recommend a five-fold dilution of the ligation mix, and adding not more than 1/10 of the diluted volume to the cells. For best results, the volume added should also not exceed 10% of the volume of the competent cells that you are using.
What are the recommended conditions for blunt-ended ligations?
Generally, ligations are done in a 20 µL volume. Use a total of 100 to 1000 ng of DNA with an insert to vector ratio of 3:1. Add 1.0 units (Weiss) ligase to the reaction. Incubate at room temperature for 4 h or overnight at 14-16 degrees C.
Ideally, assemble several reactions with varying ratios of vector:insert (i.e. 3:1, 5:1, 10:1, 20:1, etc.) to determine the optimal ratio for ligation.
Thermo Fisher Scientific offers T4 DNA ligase at two concentrations: 1 U/µL (Cat. No. 15224-017) and 5 U/µL (Cat. No. 15224-041). When performing blunt or TA cloning ligations, the higher concentration of ligase is generally preferred since ligating a blunt or single base overhang requires more enzyme.
What are the recommended conditions for cohesive-end ligations?
Generally, ligations are done in a 20 µL volume. Use a total of 10 to 100 ng of DNA per reaction with an insert to vector ratio of 3:1. Add 0.1 units (Weiss) ligase to the reaction. Incubate at room temperature for 30-60 minutes.
Optimal ligation may occur at other ratios (e.g. 1:5, 1:10). If possible, assemble several ligation reactions of varying insert to vector ratios in order to reveal the optimal ligation conditions.
Thermo Fisher Scientific offers T4 DNA ligase at two concentrations: 1 U/µL (Cat. No. 15224-017) and 5 U/µL (Cat. No. 15224-041). When performing blunt or TA cloning ligations, the higher concentration of ligase is generally preferred since ligating a blunt or single base overhang requires more enzyme.
Which is better to use, T4 or E. coli DNA ligase?
It depends on your application. For ligation of dsDNA fragments with cohesive ends, either enzyme can be used. E. coli DNA ligase requires the presence of beta-NAD, while T4 DNA ligase requires ATP. However, only T4 DNA ligase can join blunt-ended DNA fragments - E. coli ligase is unable to join such fragments.
E. coli DNA ligase is generally used to eliminate nicks during second-strand cDNA synthesis. T4 DNA ligase should not be substituted for E. coli DNA ligase in second-strand synthesis because of its capability for blunt end ligation of the ds cDNA fragments, which could result in formation of chimeric inserts.
Why is ATP present in the reaction buffer for T4 DNA Ligase?
ATP is necessary for enzymatic function. It is involved in phosphorylating the ligase prior to the ligation reaction. Ligation efficiency is markedly reduced by removing ATP from the reaction. It is important, therefore, to handle the buffer appropriately in order to minimize degradation of ATP.
What causes synthesized \peptides to be over or under the expected weight when post-cleavage mass spectral analysis is performed? What causes multiple peaks for one peptide, by RP-HPLC?
Overweight: Most often reflects failure to remove all the side-chain protecting groups from all amino acids ("incomplete cleavage"). If insufficient amounts of scavengers were used, these protecting groups may reattach, OR permanently attach to another amino acid. If this occurs, it may be necessary to modify the system used for cleavage. Increased time, better mixing, or increased scavengers may be needed. This may be best determined by trial and error.
Underweight: Usually reflects a problem during the synthesis. If the run was done with conductivity or UV monitoring during Fmoc removal, it may be possible to modify the subsequent synthesis to avoid problems at specific areas of the sequence. An analysis of the probable secondary structure of the peptide chain may also be helpful for future strategies.
How can peptide synthesis amino acid cartridges leak and spill solvents during their activation and transfer to the RV?
If these cartridges are being reused, the NMP can cause them to swell, and they no longer fit or slide well in the guideway. If the guideway or the exterior of the needles have became dirty, this can also lead to misalignment. And if you forgot to remove the metal cap, the needle cannot penetrate the septum - this may cause a spill OR stop the run.
How often should the sound of the vacuum assist be heard on the 433A?
At most, once or twice a day. If more frequent, there may be a gas leak. Nitrogen pressure is used to generate the vacuum, which assists the opening of the valves. If there is no apparent gas leak, then it is possible that a valve has failed and the solvent leakage has damaged the vacuum ballast. Both the vacuum system and the valve block need inspection.
Why doesn't the 433A manual or the "quick start card" mention the need for an extra AA cartridge in the guideway at the start of a sequence?
The newest 433A User's Manual does cover this. The barcode reader is one position ahead (left) of the needle position. The extra (empty) is needed to prevent advancement of the first cartridge until after it is read.
Why is the conductivity so high in my peptide synthesis (monitored during deprotection)?
The meter detects any ionic species. A common cause of higher than expected values is a leak of a small amount of resin from the RV into the lines and up to the in-line filters. The use of old or poor quality piperidine or NMP may also give a high background. Standard conductivity measured in micro Siemens/cm is much higher than the sensitivity of this cell. A very small amount of ionic material caused a large change in the reading. Occasionally, Fmoc amino acids have ionic contaminants which give high readings. In-line filters may also be contaminated.
What can be done when I observe errors on the 433A such as "chemistry not declared" (CND), a "PAUSE" that will not unPAUSE, a "run cannot be sent", or during a send the chirps are slower than usual?
The problem occurs when a 433A connected to a Macintosh computer interprets a signal from the computer as a "reset". Some "fixes" have been successful, particularly installation of a special optical isolator on the 433A. When the instrument has frozen in a run, the run needs to be stopped and re-started, often with a "COLD REBOOT". To prevent the problem, it is strongly suggested that no network connection be attached to the computer during a run. No other software should be run, or even loaded to the hard drive of the computer.
How can I tell whether or not detergent in a sample can be removed with Prosorb Sample Preparation Cartridges?
If the detergent identity is known, look up its CMC (critical micellar concentration); a table of these is listed in the Biological Detergents section of the Sigma-Aldrich catalog. If you are above the CMC, the detergent forms micelles that cannot be removed by the filtering membrane, so the sample must be diluted prior to application to the Prosorb Sample Preparation Cartridges.
How can I tell whether or not a reagent bottle is pressurizing correctly when using the Procise System?
You can backflush the bottle's pickup line in manual control (there is a specific function for each bottle position on the Procise System) and observe its bubbling, which should slow and then stop within a short period (depending upon how full the bottle is) as it pressurizes with argon. If it continues to bubble, either the cap assembly is leaking or the pressurizing or venting valves for the bottle are.
I am using the Procise System and have increased the final %B in the gradient to separate LYS from LEU, but now all the late PTH amino acid peaks are crushed together. What can I do?
The PTH column is worn out; you should replace it.
How can I improve peak resolution between ASP and DTT or ASN when using the Procise System?
You can move ASP (and GLU) away from DTT and to later retention times by reducing slightly the pH of Solvent A3. This is best accomplished by adding a small amount of TFA (R3)--about 50 to 100 ul/liter of Solvent A3. In the Procise System cLC, when ASP and ASN are too close together, the problem can usually be resolved by replacing the guard column (part no. 401883). Decreasing initial %B in the gradient (e.g., from 10% to 8%) will move both DTT and ASP to later retention times.
What can cause broad dips or cyclical peaks in the baseline when using the Procise System?
This can be due to pump irregularities, often caused by worn seals. A more or less regular "sawtooth" pattern is usually indicative of a failed dynamic mixer.
I want to make peptide amides, but your amide resin has no amino acid attached. Why not? Do I need to do anything special?
There is no amino acid attached because one is not needed. The amide linker has a free amine which is protected by an Fmoc group. Upon removal of the Fmoc group, an amide bond may be formed with the incoming activated amino acid. Nothing special needs to be done, although you must tell your synthesizer you are using an amide support and/or the first amino acid is not on the support. Standard cleavage protocols may be used.
How many degenerate sites are allowed in a 25-mer present at a total concentration of 1 micromolar (100 pmoles/100 mL)?
Primer mixtures with 256-fold and 32-fold degeneracies have been used [see Mack DH, Sninsky JJ (1988) Proc Natl Acad Sci USA 85:6977-6981 and Lee et al. (1988) Science 239:1288-1291.] We recommend that users synthesize pools of no more than 32- to 64-fold degenerate primers, making additional pools separately to account for all possible degeneracy. A matrix should be set up so that degeneracy is no more than 2-fold at each site, with all sites in the matrix run at the same time. Inosine may also be used for the degenerate positions in the primer. Performing touchdown PCR may help increase the specificity of degenerate primer PCR amplification.
What are some of the problems associated with sticky-end cloning?
The amplified DNA needs to be purified from the PCR mixture components prior to cloning. The dNTPs carried over from the PCR are competitive inhibitors for ATP in the ligation reaction.
If during synthesis of the PCR primers their chemical integrity has been compromised by either a base substitution or modification, the enzyme recognition site may in actuality not exist. If this is the case, PCR products will be resistant to digestion with restriction enzymes. It may be necessary to use a higher concentration of the restriction enzyme and to incubate at the appropriate temperature overnight to ensure cutting.