Ethyl 3,4,5-trihydroxybenzoate, 98%
Ethyl 3,4,5-trihydroxybenzoate, 98%
Ethyl 3,4,5-trihydroxybenzoate, 98%
Thermo Scientific Chemicals

Ethyl 3,4,5-trihydroxybenzoate, 98%

CAS: 831-61-8 | C9H10O5 | 198.174 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A13733.09
also known as A13733-09
Price (USD)
47.65
Online Exclusive
53.30
Save 5.65 (11%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
47.65
Online Exclusive
53.30
Save 5.65 (11%)
Each
Chemical Identifiers
CAS831-61-8
IUPAC Nameethyl 3,4,5-trihydroxybenzoate
Molecular FormulaC9H10O5
InChI KeyVFPFQHQNJCMNBZ-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC(O)=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream
FormPowder
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only