2-Aminophenol, 99%
2-Aminophenol, 99%
2-Aminophenol, 99%
Thermo Scientific™

2-Aminophenol, 99%

CAS: 95-55-6 | C6H7NO | 109.128 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A13735.36
also known as A13735-36
Price (USD)
102.65
Online Exclusive
114.00
Save 11.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
102.65
Online Exclusive
114.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS95-55-6
IUPAC Name2-aminophenol
Molecular FormulaC6H7NO
InChI KeyCDAWCLOXVUBKRW-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)172-177?C
Appearance (Color)White to grey to brown
Assay (GC)≥98.5%
Identification (FTIR)Conforms
2-Aminophenol is used to make dyes and pharmaceuticals. It is also used as a reagent for the synthesis of heterocyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminophenol is used to make dyes and pharmaceuticals. It is also used as a reagent for the synthesis of heterocyclic compounds.

Solubility
Soluble in water (17 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. A. Guenbour.; A. Kacemi.; A. Benbachir.; L. Aries. Electropolymerization of 2-aminophenol: Electrochemical and spectroscopic studies. Progress in Organic Coatings. 2000, 38 (2), 121-126.
  2. William P. Griffith.; Tse Yuen Koh. Vibrational spectra of 1,2-benzenedithiol, 2-aminothiophenol and 2-aminophenol and their SER spectra. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 1995, 51 (2), 18-22.