Geraniol, 97%
Geraniol, 97%
Geraniol, 97%
Thermo Scientific Chemicals

Geraniol, 97%

CAS: 106-24-1 | C10H18O | 154.253 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A13736.0B
also known as A13736-0B
Price (USD)
445.65
Online Exclusive
495.00
Save 49.35 (10%)
Each
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)
445.65
Online Exclusive
495.00
Save 49.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS106-24-1
IUPAC Name(2E)-3,7-dimethylocta-2,6-dien-1-ol
Molecular FormulaC10H18O
InChI KeyGLZPCOQZEFWAFX-JXMROGBWSA-N
SMILESCC(C)=CCC\C(C)=C\CO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4745-1.4795 @ 20?C
FormLiquid
Identification (FTIR)Conforms
Geraniol is used in the synthesis of insect repellant. It is also used in the synthesis of Angelicoin A and Herecinone J, which inhibit collagen-induced platelet aggregation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Geraniol is used in the synthesis of insect repellant. It is also used in the synthesis of Angelicoin A and Herecinone J, which inhibit collagen-induced platelet aggregation.

Solubility
Not miscible in water.

Notes
Incompatible with Strong oxidizing agents, acid chlorides, acid anhydrides. Keep away from heat. Air sensitive.
RUO – Research Use Only

General References:

  1. J. M. kim; M. R. Marshall; J.A. Cornell; J. F. Preston and C. I. wei. Antibacterial Activity of Carvacrol, Citral, and Geraniol against Salmonella typhimurium in Culture Medium and on Fish Cubes.Journal of Food Science.1995, 60 1364-1368.
  2. Yvette D. Burke; M. Jennifer Stark; Steven L. Roach; Stephanie E. Sen; Pamela L. Crowell. Inhibition of pancreatic cancer growth by the dietary isoprenoids farnesol and geraniol. Lipids.1997, 32 151-156.