Have there been any publications using the Gibco Episomal hiPSC Line?
Yes, see Burridge, P.W., Thompson, S., Millrod, M.A., et al. 2011. A universal system for highly efficient cardiac differentiation of human induced pluripotent stem cells that eliminates interline variability. PLoS One. Note that this publication refers to the Gibco Episomal hiPSC Line as clone 6.2.