Invitrogen™

Vector NTI Advance™ 1-Year Industrial License

Catalog number:  A13784

 Related applications:

Vector NTI® Software

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Vector NTI Advance® is the most complete and highly integrated application for desktop sequence analysis and molecular biology data management. Length of license: 1-year.

Its many features include a comprehensive suite of DNA and protein sequence analysis tools across five integrated modules:

• Vector NTI®—Sequence analysis, annotation, and illustration; restriction mapping; recombinant molecule design, including Gateway® and TOPO® cloning; in silico gel electrophoresis; and molecular biology data management
• AlignX®—Multiple sequence alignment of proteins and DNAs
• ContigExpress®—DNA sequence assembly and sequencing project management using the CAP 3 algorithm
• GenomBench™—Analysis and annotation of reference genomic DNA sequences
• BioAnnotator™—Functional annotation of DNAs and proteins

Vector NTI Advance® includes specific capabilities that speed and simplify experimental design:

• Robust on-board data management capabilities
• Superior graphics, readily available for export
• Non-proprietary sequence file formats
• In silico Gateway® and TOPO® cloning—Step by step cloning wizards, incompatible clonings flagged, run multiple clonings in parallel and manage cloning projects more effectively.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

License Type: Term
Operating System: Mac OS X (with Parallels Desktop 5), Mac OS X 10.5 (with Parallels Desktop 6), Mac OS X 10.6 (with Parallels Desktop 6), Windows 7, Windows Vista, Windows XP
Product Line: Vector NTI Advance®
Product Size: 1 each
Software Category: Software
Software Type: Vector NTI®
System Requirements: Mac: Intel Core i7 2.66 GHz or Intel Core 2 Duo 3.06 GHz, PC: Intel Core 2 Duo 2.33 GHz, 1 GB RAM

Contents & storage

Software CD, Installation Guide, Quick Start Guide, License Letter

Documents

Manuals & protocols