1-Iodohexane, 98%, stab. with copper
1-Iodohexane, 98%, stab. with copper
1-Iodohexane, 98%, stab. with copper
1-Iodohexane, 98%, stab. with copper
Thermo Scientific Chemicals

1-Iodohexane, 98%, stab. with copper

CAS: 638-45-9 | C6H13I | 212.074 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A13784.22
also known as A13784-22
Price (USD)
89.65
Special offer
Online exclusive
Ends: 30-Jun-2026
105.00
Save 15.35 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
89.65
Special offer
Online exclusive
Ends: 30-Jun-2026
105.00
Save 15.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS638-45-9
IUPAC Name1-iodohexane
Molecular FormulaC6H13I
InChI KeyANOOTOPTCJRUPK-UHFFFAOYSA-N
SMILESCCCCCCI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4900-1.4940 @ 20?C
FormLiquid
CommentStabilised with copper powder
Identification (FTIR)Conforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only