BG-11 Media, optimized for cyanobacteria, 6 L - FAQs
BG-11 Media, optimized for cyanobacteria, 6 L - FAQs
View additional product information for BG-11 Media, optimized for cyanobacteria - FAQs (A1379902, A1379901)
7 product FAQs found
I'm getting low basal expression of my gene of interest when using the pSyn_1 vector. What do you suggest I try?
The promoter for pSyn_1 is the Psc. Psc is a weak constitutive promoter that drives basal expression of your gene of interest. The promoter for pSyn_6, on the other hand, is the psbA promoter (PpsbA). PpsbA is a strong constitutive promoter of the psbA gene (encoding photosystem II protein D1) from Synechococcus elongatus, driving the high level expression of your gene of interest.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
What is the difference between pSyn_1 and pSyn_6?
The promoter for pSyn_1 is the Psc. Psc is a weak constitutive promoter that drives basal expression of your gene of interest. The promoter was taken from the solanesyl diphosphate synthase gene from Synechocystis sp. Strain PCC 6803. Note: Use of this weak constitutive promoter is a good fit for applications that are hindered by strong expression, such as pathway engineering or complementation of mutant genes normally expressed at low levels (Simkovsky et al., 2012).
The promoter for pSyn_6 is the psbA promoter (PpsbA). PpsbA is a strong constitutive promoter of the psbA gene (encoding photosystem II protein D1) from Synechococcus elongatus, driving high-level expression of your gene of interest. The pSyn-6 vector also includes a ribosome-binding site (GAAGGAG) for efficient initiation of translation, as well as a stop codon. A 6xHis TEV and a V5-His epitope tag is also included in the vector backbone for detection/purification of the gene of interest. NS1 (neutral site 1) homologous recombination sites are present in both vectors for the integration of the vector into the Synechococcus elongatus genome.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
What is the shelf-life/competency of Synechococcus?
After 6 months of storage, the Synechococcus did not lose their competency. We have not yet tested longer storage points.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
What is the mechanism of integration when using the Synechococcus expression kits?
A double homologous recombination event occurs between the Neutral Site 1 in the genome and between NSa and NSb in the plasmid. This results in the gene of interest and spectinomycin resistance integrating into the genome within the Neutral Site 1.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
What is the strain of Synechococcus used in your kits?
We no longer offer any strains. Our reagents and protocols were developed using Synechococcus elongatus strain PCC 7942, a model cyanobacterium.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
What is the formulation of Blue-Green-11 (BG-11) media?
We offer ready-to-use BG-11 Media, optimized for Cyanobacteria (Cat. No. A1379901). There are several BG11 media formulations. Please see our formulation below.
Our BG-11 media have been qualified for culturing Synechococcus elongatus (strain PCC 7942).
Item, Formula, (%w/v)
Boric Acid, H3BO3, 0.00287%
Manganese Chloride Tetrahydrate, MnCl2-4H2O, 0.00181%
Zinc Sulphate Heptahydrate, ZnSO4-7H2O, 0.00022%
Sodium Molybdate Dihydrate, Na2MoO4, 0.00039%
Copper Sulphate Pentahydrate, CuSO4-5H2O, 0.00008%
Sodium Nitrate, NaNO3, 0.15000%
Calcium Chloride Dihydrate, CaCl2-2H2O, 0.00270%
Ferric Ammonium Citrate (Green), C6H5 4yFexNyO7, 0.00120%
EDTA, EDTA, 0.00010%
Potassium Phosphate Dibasic, K2HPO4, 0.00390%
Magnesium Sulfate Heptahydrate, MgSO4-7H2O, 0.00750%
Sodium Carbonate Monohydrate, Na2CO3-1H2O, 0.00200%
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
What does the GeneArt Synechococcus Protein Expression Vector include?
It includes the pSyn_6 vector. This vector has been tested for protein expression in the Synechococcus elongatus PPC 7942. You can get the Synechococcus elongatus PPC 7942 from academic resources (https://genome.jgi.doe.gov/portal/synel/synel.info.html) or from the ATCC Collection (https://www.atcc.org/products/all/33912.aspx).
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.