Palmitoyl chloride, 98%
Palmitoyl chloride, 98%
Palmitoyl chloride, 98%
Thermo Scientific Chemicals

Palmitoyl chloride, 98%

CAS: 112-67-4 | C16H31ClO | 274.873 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A13812.36
also known as A13812-36
Price (USD)
193.65
Special offer
Online exclusive
Ends: 30-Jun-2026
228.00
Save 34.35 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
193.65
Special offer
Online exclusive
Ends: 30-Jun-2026
228.00
Save 34.35 (15%)
Each
Chemical Identifiers
CAS112-67-4
IUPAC Namehexadecanoyl chloride
Molecular FormulaC16H31ClO
InChI KeyARBOVOVUTSQWSS-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofA≥97.5% (U.S. specification)
Refractive Index1.4500-1.4540 @ 20°C (non-U.S. specification)
Assay (GC)≥97.5% (non-U.S. specification)
Appearance (Color)Clear, colorless to pale yellow
Palmitoyl chloride is a compound which has been used as a polymeric carrier for plasmid DNA delivery to 293T cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Palmitoyl chloride is a compound which has been used as a polymeric carrier for plasmid DNA delivery to 293T cells.

Solubility
Soluble in ether.Decomposes in water and alcohol.

Notes
Moisture Sensitive. Desiccate at room temperature. Store under inert gas. Incompatible with Strong oxidizing agents, Bases, Alcohols.
RUO – Research Use Only

General References:

  1. Haixing Cheng; Siguang Wang; Jintian Yang; Yongfeng Zhou; Deyue Yan; Haixing Cheng; Siguang Wang; Jintian Yang; Yongfeng Zhou; Deyue Yan. Synthesis and self-assembly of amphiphilic hyperbranched polyglycerols modified with palmitoyl chloride.Journal of Colloid and Interface Science.2009, 337 (1), 278-284.
  2. Gowdra K. Prakash; Kittappa M. Mahadevan. Enhancing the properties of wood through chemical modification with palmitoyl chloride.Applied Surface Science.2008, 254 1751-1756.