2-Butyne, 98%
2-Butyne, 98%
2-Butyne, 98%
Thermo Scientific Chemicals

2-Butyne, 98%

CAS: 503-17-3 | C4H6 | 54.092 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A13823.06
also known as A13823-06
Price (USD)
57.65
Online Exclusive
63.90
Save 6.25 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
57.65
Online Exclusive
63.90
Save 6.25 (10%)
Each
Chemical Identifiers
CAS503-17-3
IUPAC Namebut-2-yne
Molecular FormulaC4H6
InChI KeyXNMQEEKYCVKGBD-UHFFFAOYSA-N
SMILESCC#CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to very pale yellow
Refractive Index1.3900-1.3940 @ 20?C
FormLiquid
Assay (GC)≥97.5%
2-Butyne is used to synthesize alkylated hydroquinones in the total synthesis of Vitamin E. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Butyne is used to synthesize alkylated hydroquinones in the total synthesis of Vitamin E. It is also used as pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Avoid extremes of temperature and direct sunlight. Heat-sensitive. Avoid contact with oxidizing materials. Incompatible with Halogens. Chlorinated solvents.
RUO – Research Use Only

General References:

  1. Dongmin Shen; Lovat V.C Rees. Adsorption and diffusion of n-butane and 2-butyne in silicalite-I. Zeolites.1991, 11 684-689.
  2. Kevin C. Wallace; Andy H. Liu; William M. Davis; Richard R. Schrock. Living polymerization of 2-butyne using a well-characterized tantalum catalyst.Organometallics.1989, 8 (3), 644-654.
  3. With AlCl3, dimerizes to give the complex of the otherwise unstable tetramethylcyclobutadiene, cycloaddition of which with dimethyl acetylenedicarboxylate gives a Dewar benzene: Synthesis, 139 (1971):
  4. With a catalytic amount of AlCl3, hexamethyl Dewar benzene can be produced directly, with the 2-butyne also acting as the dienophile component: Org. Synth. Coll., 7, 256 (1990).
  5. Reacts with chlorine to give 3,4-dichloro,1,2,3,4-tetramethylcyclobutene, a precursor of various cyclobutadienyl complexes: Org. Synth. Coll., 5, 370 (1973).
  6. For use in formation of a Zr metallocycle as an intermediate in the formation of phospholes and analogous heterocycles, see Bis(cyclopentadienyl) zirconium dichloride, 12548.