1,9-Decadiyne, 97%
1,9-Decadiyne, 97%
1,9-Decadiyne, 97%
Thermo Scientific Chemicals

1,9-Decadiyne, 97%

CAS: 1720-38-3 | C10H14 | 134.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A13834.06
also known as A13834-06
Price (USD)
84.65
Special offer
Online exclusive
Ends: 30-Jun-2026
99.90
Save 15.25 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
84.65
Special offer
Online exclusive
Ends: 30-Jun-2026
99.90
Save 15.25 (15%)
Each
Chemical Identifiers
CAS1720-38-3
IUPAC Namedeca-1,9-diyne
Molecular FormulaC10H14
InChI KeyILVDYAGPHFWNQI-UHFFFAOYSA-N
SMILESC#CCCCCCCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4480-1.4530 @ 20?C
Assay (GC)≥96.0%
FormLiquid
Appearance (Color)Clear colorless to yellow

One-step synthesis of carbametacyclophane 3 from 1,9-decadiyne and diethyl acetylenedicarboxylate is described. The use of 1,9-decadiyne as a nucleophile led only to the monoalkynylation product, a selectivity that would be difficult to achieve with a catalytic in situ metalation process. 1,9-decadiyne reacts with iodobenzene (excess) to afford the bis-Sonogashira coupling product in excellent yield. 1,9-Decadiyne was dihydrated and gave 2,9-decanedione in 92% yield.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
One-step synthesis of carbametacyclophane 3 from 1,9-decadiyne and diethyl acetylenedicarboxylate is described. The use of 1,9-decadiyne as a nucleophile led only to the monoalkynylation product, a selectivity that would be difficult to achieve with a catalytic in situ metalation process. 1,9-decadiyne reacts with iodobenzene (excess) to afford the bis-Sonogashira coupling product in excellent yield. 1,9-Decadiyne was dihydrated and gave 2,9-decanedione in 92% yield.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Haruki Okawa,; Motoyoshi Sekiya,; Junichiro Osawa,; Tatsuo Wada,; Akira Yamada,; Hiroyuki Sasabe,; Toshiyuki Uryu. A Nonlinear Optical Waveguide of Poly(1,9-decadiyne). Polymer Journal. 1991, 23 147-153.
  2. Christophe Feuvrie,; Jérôme Blanchet,; Martine Bonin,; Laurent Micouin. Synthesis and Reactivity of Mixed Alkynylalanes by Direct Triethylamine-Catalyzed Alumination of Terminal Alkyne. Org. Lett.,. 2004 , 6(14),2333-2336.