FreeStyle™ F17 Expression Medium, 1 L - FAQs
FreeStyle™ F17 Expression Medium, 1 L - FAQs
View additional product information for FreeStyle™ F17 Expression Medium - FAQs (A1383503, A1383504, A1383501, A1383502)
2 product FAQs found
How long can I keep culturing FreeStyle cells after transfection?
We typically keep the culture going for 7-10 days posttransfection. After that, protein expression may drop.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the difference between FreeStyle 293 Expression Medium and FreeStyle F17 Expression Medium?
FreeStyle F17 Expression Medium was designed from a different base formulation than FreeStyle 293 Expression Medium. FreeStyle F17 medium shows equal or better performance than the FreeStye 293 medium for certain cell lines and proteins. It is capable of supporting the growth of 293 and CHO cells and offers a wider variety and selection of transient cell culture media options.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.