4,4-Diphenyl-2-butanone, 98%
4,4-Diphenyl-2-butanone, 98%
4,4-Diphenyl-2-butanone, 98%
4,4-Diphenyl-2-butanone, 98%
Thermo Scientific Chemicals

4,4-Diphenyl-2-butanone, 98%

CAS: 5409-60-9 | C16H16O | 224.303 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A13847.03
also known as A13847-03
Price (USD)
43.65
Online Exclusive
48.00
Save 4.35 (9%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
43.65
Online Exclusive
48.00
Save 4.35 (9%)
Each
Add to cart
Chemical Identifiers
CAS5409-60-9
IUPAC Name4,4-diphenylbutan-2-one
Molecular FormulaC16H16O
InChI KeyFPHXYKLKNOEKTQ-UHFFFAOYSA-N
SMILESCC(=O)CC(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or lumps
Melting Point (clear melt)44.0-51.0?C
Assay (GC)≥97.5%
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only