1,2-Phenylenediacetic acid, 98+%
1,2-Phenylenediacetic acid, 98+%
1,2-Phenylenediacetic acid, 98+%
1,2-Phenylenediacetic acid, 98+%
Thermo Scientific Chemicals

1,2-Phenylenediacetic acid, 98+%

CAS: 7500-53-0 | C10H10O4 | 194.186 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A13851.14
also known as A13851-14
Price (USD)
65.65
Online Exclusive
72.60
Save 6.95 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
65.65
Online Exclusive
72.60
Save 6.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS7500-53-0
IUPAC Name2-[2-(carboxymethyl)phenyl]acetic acid
Molecular FormulaC10H10O4
InChI KeyMMEDJBFVJUFIDD-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=CC=C1CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale brown
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)148.0-155.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only