1-Fluoro-2-nitrobenzene, 99%
1-Fluoro-2-nitrobenzene, 99%
1-Fluoro-2-nitrobenzene, 99%
Thermo Scientific Chemicals

1-Fluoro-2-nitrobenzene, 99%

CAS: 1493-27-2 | C6H4FNO2 | 141.101 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A13852.0B
also known as A13852-0B
Price (USD)
581.65
Online Exclusive
646.00
Save 64.35 (10%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
581.65
Online Exclusive
646.00
Save 64.35 (10%)
Each
Chemical Identifiers
CAS1493-27-2
IUPAC Name1-fluoro-2-nitrobenzene
Molecular FormulaC6H4FNO2
InChI KeyPWKNBLFSJAVFAB-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Assay (GC)≥98.5%
Refractive Index1.5300-1.5330 @ 20?C
Appearance (Color)Clear yellow to orange to brown
FormLiquid
1-Fluoro-2-nitrobenzene is an intermediate in the synthetic preparations of various pharmaceutical goods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoro-2-nitrobenzene is an intermediate in the synthetic preparations of various pharmaceutical goods.

Solubility
Immiscible in water.

Notes
Avoid excessive heat and light.
RUO – Research Use Only

General References:

  1. Elaine K. Fukuda; Robert T. McIverJr. Relative electron affinities of substituted benzophenones, nitrobenzenes, and quinones.J. Am. Chem. Soc.1985, 107 (8), 2291-2296.
  2. The activating effect of aryl halides by o- or p-nitro groups is most pronounced for aryl fluorides, e.g. reacts readily with arylamines in a convenient synthesis of 2-nitrodiarylamines: Synthesis, 215 (1980).