1-Bromo-4-iodobenzene, 98%
1-Bromo-4-iodobenzene, 98%
1-Bromo-4-iodobenzene, 98%
1-Bromo-4-iodobenzene, 98%
Thermo Scientific Chemicals

1-Bromo-4-iodobenzene, 98%

CAS: 589-87-7 | C6H4BrI | 282.906 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A13859.30
also known as A13859-30
Price (USD)
720.65
Online Exclusive
801.00
Save 80.35 (10%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
720.65
Online Exclusive
801.00
Save 80.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS589-87-7
IUPAC Name1-bromo-4-iodobenzene
Molecular FormulaC6H4BrI
InChI KeyUCCUXODGPMAHRL-UHFFFAOYSA-N
SMILESBrC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pink to brown
Identification (FTIR)Conforms
Assay (GC)≥97.5%
FormCrystalline powder
Melting Point (clear melt)89-93?C
It is mainly used as the OLED pharmaceutical intermediate, as reagent for in situ desilylation and coupling of silylated alkynes, as substrate in copper-free Sonogashira coupling in aqueous acetone. It is also used in synthesis of β,β,dibromostyrenes, as starting reagent in the total syntheses of ent-conduramine A and ent-7-deoxypancratistatin (alkaloids).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is mainly used as the OLED pharmaceutical intermediate, as reagent for in situ desilylation and coupling of silylated alkynes, as substrate in copper-free Sonogashira coupling in aqueous acetone. It is also used in synthesis of β,β,dibromostyrenes, as starting reagent in the total syntheses of ent-conduramine A and ent-7-deoxypancratistatin (alkaloids).

Solubility
Insoluble in water.

Notes
Light sensitive. Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, light.
RUO – Research Use Only

General References:

  1. Shenlin Huang; James M. Tour. Rapid bi-directional synthesis of oligo(1, 4-phenylene ethynylene)s. Tetrahedron Letters. 1999, 40 (17), 3447-3450.
  2. Harry A.J. Oonk; Peter R. van der Linde; Jan Huinink; Jacobus G. Blok. Representation and assessment of vapour pressure data; a novel approach applied to crystalline 1-bromo-4-chlorobenzene, 1-chloro-4-iodobenzene, and 1-bromo-4-iodobenzene. J. Chem. Thermodyn. 1998, 30 (7), 897-907.