1,5-Dinitronaphthalene, 97+%
1,5-Dinitronaphthalene, 97+%
1,5-Dinitronaphthalene, 97+%
1,5-Dinitronaphthalene, 97+%
Thermo Scientific Chemicals

1,5-Dinitronaphthalene, 97+%

CAS: 605-71-0 | C10H6N2O4 | 218.168 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A13869.14
also known as A13869-14
Price (USD)
29.40
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
29.40
Each
Add to cart
Chemical Identifiers
CAS605-71-0
IUPAC Name1,5-dinitronaphthalene
Molecular FormulaC10H6N2O4
InChI KeyZUTCJXFCHHDFJS-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC2=C(C=CC=C12)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow or cream to pale brown
Melting Point (clear melt)212.0-222.0°C
Assay (GC)≥97.0%
FormCrystals or powder or crystalline powder
CommentMay contain a small amount of dark particulate matter from the manufacturing process
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only