4-Hydrazinobenzoic acid, 97%
4-Hydrazinobenzoic acid, 97%
4-Hydrazinobenzoic acid, 97%
4-Hydrazinobenzoic acid, 97%
Thermo Scientific Chemicals

4-Hydrazinobenzoic acid, 97%

CAS: 619-67-0 | C7H8N2O2 | 152.153 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A13874.30
also known as A13874-30
Price (USD)
606.65
Special offer
Online exclusive
Ends: 30-Jun-2026
714.00
Save 107.35 (15%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
606.65
Special offer
Online exclusive
Ends: 30-Jun-2026
714.00
Save 107.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS619-67-0
IUPAC Name4-hydrazinylbenzoic acid
Molecular FormulaC7H8N2O2
InChI KeyPCNFLKVWBDNNOW-UHFFFAOYSA-N
SMILESNNC1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to orange to pale brown
FormPowder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (HPLC)≥96.0%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only