N,N'-Bis(salicylidene)ethylenediamine, 98%
N,N'-Bis(salicylidene)ethylenediamine, 98%
N,N'-Bis(salicylidene)ethylenediamine, 98%
N,N'-Bis(salicylidene)ethylenediamine, 98%
Thermo Scientific Chemicals

N,N'-Bis(salicylidene)ethylenediamine, 98%

CAS: 94-93-9 | C16H16N2O2 | 268.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A13885.22
also known as A13885-22
Price (USD)
177.65
Online Exclusive
197.00
Save 19.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
177.65
Online Exclusive
197.00
Save 19.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS94-93-9
IUPAC Name(6Z)-6-({[2-({[(1Z)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}amino)ethyl]amino}methylidene)cyclohexa-2,4-dien-1-one
Molecular FormulaC16H16N2O2
InChI KeyRQHVNNWVDLRULK-XSYHWHKQSA-N
SMILESO=C1C=CC=C\C1=C\NCCN\C=C1\C=CC=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤1.0%
Appearance (Color)Yellow
FormCrystalline powder or powder and/or flakes
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (GC)≥97.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only