4-Hydroxy-3,5-dimethoxybenzyl alcohol, 97%
4-Hydroxy-3,5-dimethoxybenzyl alcohol, 97%
4-Hydroxy-3,5-dimethoxybenzyl alcohol, 97%
Thermo Scientific Chemicals

4-Hydroxy-3,5-dimethoxybenzyl alcohol, 97%

CAS: 530-56-3 | C9H12O4 | 184.191 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A13888.06
also known as A13888-06
Price (USD)
79.65
Special offer
Online exclusive
Ends: 28-Dec-2026
93.20
Save 13.55
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
79.65
Special offer
Online exclusive
Ends: 28-Dec-2026
93.20
Save 13.55
Each
Chemical Identifiers
CAS530-56-3
IUPAC Name4-(hydroxymethyl)-2,6-dimethoxyphenol
Molecular FormulaC9H12O4
InChI KeyLUOAEJWSKPQLJD-UHFFFAOYSA-N
SMILESCOC1=CC(CO)=CC(OC)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale brown
Melting Point (clear melt)127.5-136.5?C
Assay (Silylated GC)≥96.0%
FormPowder
Identification (FTIR)Conforms
4-Hydroxy-3,5-dimethoxybenzyl alcohol is a 2,4,6- substituted phenol with antiviral properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-3,5-dimethoxybenzyl alcohol is a 2,4,6- substituted phenol with antiviral properties.

Solubility
Sparingly soluble (0.40 g/L at 25°C).

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
RUO – Research Use Only