3,4-Dichlorophenylhydrazine hydrochloride, 98+%
3,4-Dichlorophenylhydrazine hydrochloride, 98+%
3,4-Dichlorophenylhydrazine hydrochloride, 98+%
3,4-Dichlorophenylhydrazine hydrochloride, 98+%
Thermo Scientific Chemicals

3,4-Dichlorophenylhydrazine hydrochloride, 98+%

CAS: 19763-90-7 | C6H7Cl3N2 | 213.486 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A13906.09
also known as A13906-09
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
79.90
Save 12.25 (15%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
79.90
Save 12.25 (15%)
Each
Add to cart
Chemical Identifiers
CAS19763-90-7
IUPAC Namehydrogen (3,4-dichlorophenyl)hydrazine chloride
Molecular FormulaC6H7Cl3N2
InChI KeyWIKZRYSGQPZTMM-UHFFFAOYSA-N
SMILES[H+].[Cl-].NNC1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale red
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only