3-Chlorobenzoylacetonitrile, 98%
3-Chlorobenzoylacetonitrile, 98%
3-Chlorobenzoylacetonitrile, 98%
Thermo Scientific Chemicals

3-Chlorobenzoylacetonitrile, 98%

CAS: 21667-62-9 | C9H6ClNO | 179.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A13913.03
also known as A13913-03
Price (USD)
25.65
Online Exclusive
28.40
Save 2.75 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
25.65
Online Exclusive
28.40
Save 2.75 (10%)
Each
Chemical Identifiers
CAS21667-62-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale orange
Assay (GC)≥97.5%
Melting Point (clear melt)79.5-85.5?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only