Bromoacetonitrile, 96%
Bromoacetonitrile, 96%
Bromoacetonitrile, 96%
Thermo Scientific Chemicals

Bromoacetonitrile, 96%

CAS: 590-17-0 | C2H2BrN | 119.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A13933.30
also known as A13933-30
Price (USD)
483.65
Online Exclusive
537.00
Save 53.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
483.65
Online Exclusive
537.00
Save 53.35 (10%)
Each
Chemical Identifiers
CAS590-17-0
IUPAC Name2-bromoacetonitrile
Molecular FormulaC2H2BrN
InChI KeyREXUYBKPWIPONM-UHFFFAOYSA-N
SMILESBrCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0% (non-U.S. Specification)
Assay from Suppliers CofA≥95.0% (U.S. specification)
CommentPurchased in the U.S. and in other countries
Identification (FTIR)Conforms (Non-U.S. specification)
Appearance (Color)Clear colorless to yellow
View more
Bromoacetonitrile was used in the synthesis of 1-cyanomethyl-1,1-dimethylhydrazinium bromide. It is used as an alkalyting agent for the nitrogen phosphorus detector.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoacetonitrile was used in the synthesis of 1-cyanomethyl-1,1-dimethylhydrazinium bromide. It is used as an alkalyting agent for the nitrogen phosphorus detector.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.
RUO – Research Use Only

General References:

  1. Carles Miró Sabaté; Henri Delalu; Erwann Jeanneau. Synthesis, characterization, and energetic properties of salts of the 1-cyanomethyl-1,1-dimethylhydrazinium cation. Chemistry An Asian Journal. 2012, 7 (5), 1085-1095 .
  2. Ho-Sang. Shin; Seungki. Kim; Seung-Woon. Myung; Jong-Sei. Park. Iodoacetonitrile and Bromoacetonitrile as Alkylating Reagents for the Nitrogen-Phosphorus Detector. Anal. Chem. 1995, 67 (11), 1853-1859.
  3. Phenols can be protected (K2CO3, acetone) as their cyanomethyl ethers which can be cleaved by hydrogenation of the nitrile: Tetrahedron Lett., 34, 7567 (1993).
  4. The Reformatsky-type reaction with cyclohexanone is more successful in the presence of a Zn/Ag couple than with the conventional Zn-Cu couple: Synth. Commun., 19, 2355 (1989). Good yields of ß-hydroxy nitriles have also been achieved from aldehydes and ketones using Zn and TMS chloride: Tetrahedron Lett., 31, 2205 (1990). The condensation with aldehydes can also be mediated by active Ni: Tetrahedron Lett., 26, 155 (1985). The reaction with aldehydes in the presence of Zn and tri-n-butylphosphine gives ɑß-unsaturated nitriles directly: Synth. Commun., 20, 3277 (1990).