2-Methylcyclopentane-1,3-dione, 98%
2-Methylcyclopentane-1,3-dione, 98%
2-Methylcyclopentane-1,3-dione, 98%
Thermo Scientific Chemicals

2-Methylcyclopentane-1,3-dione, 98%

CAS: 765-69-5 | C6H8O2 | 112.128 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A13936.14
also known as A13936-14
Price (USD)
129.65
Online Exclusive
144.00
Save 14.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
129.65
Online Exclusive
144.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS765-69-5
IUPAC Name2-methylcyclopentane-1,3-dione
Molecular FormulaC6H8O2
InChI KeyHXZILEQYFQYQCE-UHFFFAOYSA-N
SMILESCC1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals
Identification (FTIR)Conforms
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)210-220?C
Appearance (Color)White to pale brown
View more
2-Methylcyclopentane-1,3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone. The alkylation of 2-methylcyclopentane-1,3-dione (A) with 2-(1-naphthyl)ethyl bromide gives only the O-alkylation product. Mixtures of C- and O-alkyl derivatives are obtained. Michael addition of 2-methylcyclopentane-1,3-dione to methyl vinyl ketone in water gives 2-methyl-2-(3-oxo- butyl)-1,3-cyclopentanedione (3), an important intermediate of natural product chemistry. 2-Methylcyclopentane-1, 3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclopentane-1,3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone. The alkylation of 2-methylcyclopentane-1,3-dione (A) with 2-(1-naphthyl)ethyl bromide gives only the O-alkylation product. Mixtures of C- and O-alkyl derivatives are obtained. Michael addition of 2-methylcyclopentane-1,3-dione to methyl vinyl ketone in water gives 2-methyl-2-(3-oxo- butyl)-1,3-cyclopentanedione (3), an important intermediate of natural product chemistry. 2-Methylcyclopentane-1, 3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone.

Solubility
Solubility in hot Methanol (almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Joseph P. John,; S. Swaminathan,; P. S. Venkataramani. 2-Methylcyclopentane-1,3-Dione. Organic Syntheses. year of publication , volume issue , pg no.
  2. Victor J. Grenda,; Glenn W. Lindberg,; Norman L. Wendler,; Seemon H. Pines. A convenient synthesis of 2-methylcyclopentane-1,3-dione . J. Org. Chem. 1967, 32 (4), 1236-1237.
  3. Valuable intermediate for steroid synthesis. For a review, see: Synthesis, 477 (1989). Condensation with methyl vinyl ketone and proline-catalyzed asymmetric aldol cyclization of the product gives a diketone useful in steroid and prostaglandin synthesis: Org. Synth. Coll., 7, 363 (1990):