p-Xylylene dichloride is a monomer used in preparation of poly(p-phenylene vinylene(PPV) nanotubes and nanorods by chemical vapor deposition (CVD) polymerization method. It was used in a study to investigate the effect of CVD monomer selection and reaction conditions on the resulting PPV polymer layer composition and luminescence properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
p-Xylylene dichloride is a monomer used in preparation of poly(p-phenylene vinylene(PPV) nanotubes and nanorods by chemical vapor deposition (CVD) polymerization method. It was used in a study to investigate the effect of CVD monomer selection and reaction conditions on the resulting PPV polymer layer composition and luminescence properties.
Solubility
Insoluble in water
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases and active metals.
RUO – Research Use Only
General References:
- Kai Yu Wang; Qian Yang; Tai-Shung Chung; Raj Rajagopalan. Enhanced forward osmosis from chemically modified polybenzimidazole (PBI) nanofiltration hollow fiber membranes with a thin wall. Chemical Engineering Science. 2009, 64,(7), 1577-1584.
- M.P.Tsyurupa; V.A.Davankov. Hypercrosslinked polymers: basic principle of preparing the new class of polymeric materials. Reactive and Functional Polymers. 2002, 53,(2-3), 193-203.