Pyrazine, 99+% (dry wt.) water <1.0%
Pyrazine, 99+% (dry wt.) water <1.0%
Pyrazine, 99+% (dry wt.) water <1.0%
Thermo Scientific Chemicals

Pyrazine, 99+% (dry wt.) water <1.0%

CAS: 290-37-9 | C4H4N2 | 80.09 g/mol
Have Questions?
Catalog NumberQuantity
A13982.14
also known as A13982-14
25 g
Catalog number A13982.14
also known as A13982-14
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
71.00
Save 10.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
71.00
Save 10.35 (15%)
Each
Chemical Identifiers
CAS290-37-9
IUPAC Namepyrazine
Molecular FormulaC4H4N2
InChI KeyKYQCOXFCLRTKLS-UHFFFAOYSA-N
SMILESC1=CN=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥99.0% (dry wt. basis)
FormCrystals or powder or crystalline powder or fused solid
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤1%
Pyrazine is used in flavoring, fragrance, and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrazine is used in flavoring, fragrance, and dyes.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Lainne E.Seitz; William J.Suling; Robert C.Reynolds. Synthesis and antimycobacterial activity of pyrazine and quinoxaline derivatives. J. Med. Chem. 2002, 45,(25), 5604-5606.
  2. A.Raab; G.A.Worth; H.D.Meyer; L.S.Cederbaum. Molecular dynamics of pyrazine after excitation to the S2 electronic state using a realistic 24-mode model Hamiltonian. J. Chem. Phys. 1999, 110,(2), 936.