Ethyl hexanoate, 99%
Ethyl hexanoate, 99%
Ethyl hexanoate, 99%
Ethyl hexanoate, 99%
Thermo Scientific Chemicals

Ethyl hexanoate, 99%

CAS: 123-66-0 | C8H16O2 | 144.214 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A13985.36
also known as A13985-36
Price (USD)
75.65
Online Exclusive
84.20
Save 8.55 (10%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
75.65
Online Exclusive
84.20
Save 8.55 (10%)
Each
Add to cart
Chemical Identifiers
CAS123-66-0
IUPAC Nameethyl hexanoate
Molecular FormulaC8H16O2
InChI KeySHZIWNPUGXLXDT-UHFFFAOYSA-N
SMILESCCCCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4060-1.4090 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
FormLiquid
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only