Copper(II) sulfate, anhydrous, 98%
Copper(II) sulfate, anhydrous, 98%
Copper(II) sulfate, anhydrous, 98%
Copper(II) sulfate, anhydrous, 98%
Thermo Scientific Chemicals

Copper(II) sulfate, anhydrous, 98%

Copper(II) sulfate, 98%, CuO4S, CAS Number-7758-98-7, copper 2+ sulfate, copper ii sulfate, blue stone, copper monosulfate, copper sulphate, trinagle, copper sulfate, hylinec, cupric sulfate anhydrous, copper ii sulfate, anhydrous, 1000g, CHEBI:23414, 3.6, Hazard Class: 9; Packaging Group: III | CAS: 7758-98-7 | CuO4S | 159.602 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A13986.0B
Price (USD)
246.65
Online Exclusive
274.00
Save 27.35 (10%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
246.65
Online Exclusive
274.00
Save 27.35 (10%)
Each
Chemical Identifiers
CAS7758-98-7
IUPAC Namecopper(2+) sulfate
Molecular FormulaCuO4S
InChI KeyARUVKPQLZAKDPS-UHFFFAOYSA-L
SMILES[Cu++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Elemental AnalysisZinc (Zn): < 300ppm
Appearance (Color)White to pale grey
Assay from Supplier's CofA≥97.5%
Elemental AnalysisLead (Pb): < 25ppm
View more
Copper(II)sulfate is used in electroplating and in mining industries. It is used as a dehydrating agent for forming and manipulating acetal groups. In analytical chemistry, it is used in Fehling's solution and Benedict's solution to test for reducing sugars. Also, used to test protein in Biuret test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II)sulfate is used in electroplating and in mining industries. It is used as a dehydrating agent for forming and manipulating acetal groups. In analytical chemistry, it is used in Fehling′s solution and Benedict′s solution to test for reducing sugars. Also, used to test protein in Biuret test.

Solubility
Soluble in water and methanol. Insoluble in ethanol.

Notes
Incompatible with strong bases, metals, alkali metals and powdered metals.
RUO – Research Use Only

General References:

  1. Dai, Q.; Zhu, X.; Abbas, S.; Karangwa, E.; Zhang, X.; Xia, S.; Feng, B.; Jia, C. Stable Nanoparticles Prepared by Heating Electrostatic Complexes of Whey Protein Isolate-Dextran Conjugate and Chondroitin Sulfate. J. Agric. Food Chem. 2015, 63 (16), 4179-4189.
  2. Lu, D.; Hossain, D.; Jia, Z.; Monteiro, M. J. One-Pot Orthogonal Copper-Catalyzed Synthesis and Self-Assembly of L-Lysine-Decorated Polymeric Dendrimers. Macromolecules 2015, 48 (6), 1688-1702.