4'-Bromo-3-chloropropiophenone, 94%
4'-Bromo-3-chloropropiophenone, 94%
4'-Bromo-3-chloropropiophenone, 94%
4'-Bromo-3-chloropropiophenone, 94%
Thermo Scientific Chemicals

4'-Bromo-3-chloropropiophenone, 94%

CAS: 31736-73-9 | C9H8BrClO | 247.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A13988.30
also known as A13988-30
Price (USD)
839.65
Online Exclusive
933.00
Save 93.35 (10%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
839.65
Online Exclusive
933.00
Save 93.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS31736-73-9
IUPAC Name1-(4-bromophenyl)-3-chloropropan-1-one
Molecular FormulaC9H8BrClO
InChI KeySYWQQEVHZPPLNQ-UHFFFAOYSA-N
SMILESClCCC(=O)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)Cream to pale yellow
Melting Point (clear melt)54.0-66.0?C
Assay (GC)≥92.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only