10-Undecen-1-ol is used as a flavoring agent. It reacts with tosylated beta-cyclodextrin to prepare beta-cyclodextrin undecenylether. It is used in the synthesis of 10-Undecenylacrylat by reacting with acryloyl and triethylamine. It reacts with carbon monoxide in the presence of dicobalt octacarbonyl and pyridine as a base at pressures greater than 150 bar and temperatures in the range of 100 to 2000C to linear and branched polyesters having relatively high molecular weights (Mn> 10,000 g / mol) in good yields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
10-Undecen-1-ol is used as a flavoring agent. It reacts with tosylated beta-cyclodextrin to prepare beta-cyclodextrin undecenylether. It is used in the synthesis of 10-Undecenylacrylat by reacting with acryloyl and triethylamine. It reacts with carbon monoxide in the presence of dicobalt octacarbonyl and pyridine as a base at pressures greater than 150 bar and temperatures in the range of 100 to 2000C to linear and branched polyesters having relatively high molecular weights (Mn> 10,000 g / mol) in good yields.
Solubility
Miscible with alcohol and ether. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
- Useful building block, for example for long-chain ω-unsaturated acids: J. Org. Chem., 54, 1948 (1989).
- Legostaeva, Y. V.; Botsman, L. P.; Nazarov, I. S.; Yakovleva, M. P.; Garifullina, L. R.; Khalikov, R. M.; Ishmuratov, G. Y. One-pot ozonolytic synthesis of acyclic alpha,μ-bifunctional compounds from methyl 10-undecenoate and 10-undecen-1-ol. Russ. J. Appl. Chem. 2015, 88 (6), 935-940.
- Hou, L.; Wang, W.; Sheng, J.; Liu, C. Synthesis and 13C-NMR analysis of an ethylene copolymer with 9-decen-1-ol. RSC Adv. 2015, 5 (120), 98929-98933.