Copper(II) nitrate hemi(pentahydrate), 98%
Copper(II) nitrate hemi(pentahydrate), 98%
Copper(II) nitrate hemi(pentahydrate), 98%
Thermo Scientific Chemicals

Copper(II) nitrate hemi(pentahydrate), 98%

CAS: 19004-19-4 | CuN2O6·2.5H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A14004.30
also known as A14004-30
Price (USD)
52.60
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
52.60
Each
Chemical Identifiers
CAS19004-19-4
IUPAC Namedicopper(2+) pentahydrate tetranitrate
Molecular FormulaCu2H10N4O17
InChI KeyIZDJJEMZQZQQQQ-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or lumpy powder
Residual water17.8-22.4% Water
Assay (unspecified)>97.5%
Appearance (Color)Blue
Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.

Solubility
Soluble in water, ammonia and ethanol

Notes
Incompatible with reducing agents, organic materials and powdered metals.
RUO – Research Use Only

General References:

  1. Lu, Y.; Chen, J.; Wang, A.; Bao, N.; Feng, J.; Wang, W.; Shao, L. Facile synthesis of oxygen and sulfur co-doped graphitic carbon nitride fluorescent quantum dots and their application for mercury(II) detection and bioimaging. J. Mater. Chem. C 2015, 3 (1), 73-78.
  2. Dichiara, A. B.; Webber, M. R.; Gorman, W. R.; Rogers, R. E. Removal of Copper Ions from Aqueous Solutions via Adsorption on Carbon Nanocomposites. ACS Appl. Mater. Interfaces 2015, 7 (28), 15674-15680.