Methyl 2-furoate, 98+%
Methyl 2-furoate, 98+%
Methyl 2-furoate, 98+%
Thermo Scientific Chemicals

Methyl 2-furoate, 98+%

CAS: 611-13-2 | C6H6O3 | 126.111 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A14009.14
also known as A14009-14
Price (USD)
31.70
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
31.70
Each
Chemical Identifiers
CAS611-13-2
IUPAC Namemethyl furan-2-carboxylate
Molecular FormulaC6H6O3
InChI KeyHDJLSECJEQSPKW-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.4830-1.4900 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only