Sodium phenylpyruvate monohydrate, 98%
Sodium phenylpyruvate monohydrate, 98%
Sodium phenylpyruvate monohydrate, 98%
Sodium phenylpyruvate monohydrate, 98%
Thermo Scientific Chemicals

Sodium phenylpyruvate monohydrate, 98%

CAS: 122049-54-1 | C9H7O3 | 163.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
50 g
Catalog number A14027.06
also known as A14027-06
Price (USD)
48.65
Online Exclusive
53.70
Save 5.05 (9%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
48.65
Online Exclusive
53.70
Save 5.05 (9%)
Each
Add to cart
Chemical Identifiers
CAS122049-54-1
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to pale cream or pale yellow
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)4.4-13.2% (0.5-1.5 waters)
Sodium phenylpyruvate monohydrate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phenylpyruvate monohydrate is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Zhiwei Guo; Animesh Goswami; K.David Mirfakhrae; Ramesh N.Patel. Asymmetric acyloin condensation catalyzed by phenylpyruvate decarboxylase. Tetrahedron: Asymmetry. 1999, 10,(24), 4667-4675.
  2. Jeffrey D.Hermes; Paul M.Weiss; W.W.Cleland. Use of nitrogen-15 and deuterium isotope effects to determine the chemical mechanism of phenylalanine ammonia-lyase. Biochemistry. 1985, 24,(12), 2959-2967.