Anthracene-9-carboxylic acid, 98+%
Anthracene-9-carboxylic acid, 98+%
Anthracene-9-carboxylic acid, 98+%
Anthracene-9-carboxylic acid, 98+%
Thermo Scientific Chemicals

Anthracene-9-carboxylic acid, 98+%

CAS: 723-62-6 | C15H10O2 | 222.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A14049.30
also known as A14049-30
Price (USD)
905.65
Online Exclusive
1,006.00
Save 100.35 (10%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
905.65
Online Exclusive
1,006.00
Save 100.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS723-62-6
IUPAC Nameanthracene-9-carboxylic acid
Molecular FormulaC15H10O2
InChI KeyXGWFJBFNAQHLEF-UHFFFAOYSA-N
SMILESOC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-green
FormPowder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Clarity0.5% w/v in ethanol: clear solution
Anthracene-9-carboxylic acid is a Cl- transport inhibitor with a moderate to strong inhibitory action on PKA activated cardiac IcI.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anthracene-9-carboxylic acid is a Cl- transport inhibitor with a moderate to strong inhibitory action on PKA activated cardiac IcI.

Solubility
Soluble to 10 mm in ethanol and to 100 mm in DMSO. Soluble in benzene, chloroform, and hydronaphthalenes. Insoluble in water.

Notes
Store in room temperature. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. H.A. Azab; S.A. El-Korashy; Z.M. Anwar; B.H.M. Hussein; G.M. Khairy. Synthesis and fluorescence properties of Eu-anthracene-9-carboxylic acid towards N-acetyl amino acids and nucleotides in different solvents. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2010, 75(1), 21-27.
  2. Michael J. Welsh. Anthracene-9-carboxylic acid inhibits an apical membrane, chloride conductance in canine tracheal epithelium. The Journal of Membrane Biology. 1984, 78(1), 61-71.