1-Bromo-3,5-difluorobenzene, 98%
1-Bromo-3,5-difluorobenzene, 98%
1-Bromo-3,5-difluorobenzene, 98%
Thermo Scientific Chemicals

1-Bromo-3,5-difluorobenzene, 98%

CAS: 461-96-1 | C6H3BrF2 | 192.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
250 g
Catalog number A14053.30
also known as A14053-30
Price (USD)
734.65
Special offer
Online exclusive
Ends: 30-Jun-2026
864.00
Save 129.35 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
734.65
Special offer
Online exclusive
Ends: 30-Jun-2026
864.00
Save 129.35 (15%)
Each
Chemical Identifiers
CAS461-96-1
IUPAC Name1-bromo-3,5-difluorobenzene
Molecular FormulaC6H3BrF2
InChI KeyJHLKSIOJYMGSMB-UHFFFAOYSA-N
SMILESFC1=CC(Br)=CC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4970-1.5010 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only