2-Chlorobenzenesulfonyl chloride, 98%
2-Chlorobenzenesulfonyl chloride, 98%
2-Chlorobenzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

2-Chlorobenzenesulfonyl chloride, 98%

CAS: 2905-23-9 | C6H4Cl2O2S | 211.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A14054.14
also known as A14054-14
Price (USD)
268.65
Online Exclusive
298.00
Save 29.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
268.65
Online Exclusive
298.00
Save 29.35 (10%)
Each
Chemical Identifiers
CAS2905-23-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless/translucent to yellow or orange or pink
FormLow melting crystals or powder or crystalline powder or clear liquid
Assay (GC)≥97.5%
Refractive Index1.5740-1.5780 @ 20?C (as melt)
2-Chlorobenzenesulfonyl chloride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzenesulfonyl chloride is used as an organic chemical synthesis intermediate.

Solubility
It reacts with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. V.N.Petrova; V.M.Petrov; G.V.Girichev; H.Oberhammer; S.N.Ivanov. Electron diffraction and quantum-chemical study of the molecular structure of 2-chlorobenzenesulfonyl chloride. Journal of Structural Chemistry. 2008, 49,(5), 850-858. 
  2. G.D.Sokolova; P.S.Khokhlov. Synthesis of N-(2-chlorophenylsulfonyl)-N'-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)-urea with a radioactive label. Chemistry of Heterocyclic Compounds. 1989, 25,(5), 547-550.