Antimony(III) fluoride, 98%
Antimony(III) fluoride, 98%
Antimony(III) fluoride, 98%
Thermo Scientific Chemicals

Antimony(III) fluoride, 98%

CAS: 7783-56-4 | F3Sb | 178.755 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A14068.30
also known as A14068-30
Price (USD)
99.65
Online Exclusive
111.00
Save 11.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
99.65
Online Exclusive
111.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS7783-56-4
IUPAC Nametrifluorostibane
Molecular FormulaF3Sb
InChI KeyGUNJVIDCYZYFGV-UHFFFAOYSA-K
SMILESF[Sb](F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale grey
Formpowder or lumps
Assay (unspecified)> 97.5% by Floride EA
Elemental AnalysisTotal fluoride > 31.09%
Antimony(III) fluoride is used as a reagent in inorganic and organofluorine chemistry. It acts as a fluorination reagent and electroplating agent. Further, it is used in the preparation of dye, pottery, porcelains, ceramic enamels and glazes. It is utilized in the preparation of freon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antimony(III) fluoride is used as a reagent in inorganic and organofluorine chemistry. It acts as a fluorination reagent and electroplating agent. Further, it is used in the preparation of dye, pottery, porcelains, ceramic enamels and glazes. It is utilized in the preparation of freon.

Solubility
Soluble in water, methanol and acetone. Insoluble in ammonia.

Notes
Moisture sensitive. It is incompatible with acids, perchloric acid, water and oxidizing agents.
RUO – Research Use Only

General References:

  1. Braunschweig, H.; Dewhurst, R. D.; Hupp, F.; Wolf, J. Unprecedented Oxidative Addition and Metal-Only Lewis Pair Chemistry of Antimony Trihalides. Chem. Eur. J. 2015, 21 (5), 1860-1862.
  2. Zemnukhova, L. A.; Udovenko, A. A.; Makarenko, N. V.; Fedorishcheva, G. A.; Polyantsev, M. M.; Kavun, V. Y. Synthesis, crystal structure, and properties of pentadecafluorotetraantimonate(III) CsRb2Sb4F15. J. Fluorine Chem. 2015, 178, 131-135.