3,5-Diaminobenzoic acid, 96%, may contain up to 3% moisture
3,5-Diaminobenzoic acid, 96%, may contain up to 3% moisture
3,5-Diaminobenzoic acid, 96%, may contain up to 3% moisture
Thermo Scientific Chemicals

3,5-Diaminobenzoic acid, 96%, may contain up to 3% moisture

CAS: 535-87-5 | C7H8N2O2 | 152.153 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
500 g
Catalog number A14075.18
also known as A14075-18
Price (USD)
31.65
Online Exclusive
35.40
Save 3.75 (11%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
31.65
Online Exclusive
35.40
Save 3.75 (11%)
Each
Chemical Identifiers
CAS535-87-5
IUPAC Name5-carboxybenzene-1,3-bis(aminium) dichloride
Molecular FormulaC7H10Cl2N2O2
InChI KeyIGPLRBRBTUNCRT-UHFFFAOYSA-N
SMILES[Cl-].[Cl-].[NH3+]C1=CC(=CC([NH3+])=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0% to ≤104.0% (dry wt. basis)
Assay (Silylated GC)≥96.0%
Appearance (Color)Grey or brown
Water Content (Karl Fischer Titration)≤3.5%
3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.

Solubility
Very soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. A.L.Ahmad; B.S.Ooi; J.P.Choudhury. Preparation and characterization of co-polyamide thin film composite membrane from piperazine and 3,5-diaminobenzoic acid. Desalination. 2003, 158,(1-3), 101-108.
  2. Hans R.Kricheldorf; Gerd Lohden. New polymer syntheses, 79. Hyperbranched poly(ester-amide)s based on 3-hydroxybenzoic acid and 3,5-diaminobenzoic acid. Macromolecular Chemistry and Physics. 1995, 196,(6), 1839-1854.