3-Chlorobenzylamine, 97%
3-Chlorobenzylamine, 97%
3-Chlorobenzylamine, 97%
Thermo Scientific Chemicals

3-Chlorobenzylamine, 97%

CAS: 4152-90-3 | C7H8ClN | 141.598 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A14078.09
also known as A14078-09
Price (USD)
77.65
Online exclusive
86.10
Save 8.45
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
77.65
Online exclusive
86.10
Save 8.45
Each
Chemical Identifiers
CAS4152-90-3
IUPAC Name1-(3-chlorophenyl)methanamine
Molecular FormulaC7H8ClN
InChI KeyBJFPYGGTDAYECS-UHFFFAOYSA-N
SMILESNCC1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥96.0%
Refractive Index1.5565-1.5615 @ 20°C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only