4-Aminothiophenol, 97%
4-Aminothiophenol, 97%
4-Aminothiophenol, 97%
4-Aminothiophenol, 97%
Thermo Scientific Chemicals

4-Aminothiophenol, 97%

CAS: 1193-02-8 | C6H6NS | 124.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14082.22
also known as A14082-22
Price (USD)
563.65
Special offer
Online exclusive
Ends: 30-Jun-2026
663.00
Save 99.35 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
563.65
Special offer
Online exclusive
Ends: 30-Jun-2026
663.00
Save 99.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS1193-02-8
IUPAC Name(4-aminophenyl)sulfanide
Molecular FormulaC6H6NS
InChI KeyWCDSVWRUXWCYFN-UHFFFAOYSA-M
SMILESNC1=CC=C([S-])C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormFused solid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)33.0-43.0?C
Appearance (Color)Yellow
4-Aminothiophenol is used in the synthesis of pectin-4-aminothiophenol conjugates and it forms self-assembled monolayers (SAMs) in the development of DNA. It is used in the detection of thrombin by protein recognition using surface-enhanced Raman spectroscopy (SERS), which finds applications in silver and gold nano particles. It acts as a linker to thiol (SH) functionalizes multiwalled carbon nanotubes (MWCNTs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminothiophenol is used in the synthesis of pectin-4-aminothiophenol conjugates and it forms self-assembled monolayers (SAMs) in the development of DNA. It is used in the detection of thrombin by protein recognition using surface-enhanced Raman spectroscopy (SERS), which finds applications in silver and gold nano particles. It acts as a linker to thiol (SH) functionalizes multiwalled carbon nanotubes (MWCNTs).

Solubility
Immiscible with water.

Notes
Air sensitive. Store in a cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Cao, B.; Liu, B.; Yang, J. Facile synthesis of single crystalline gold nanoplates and SERS investigations of 4-aminothiophenol. Cryst. Eng. Comm. 2013, 15, 5735-5738.
  2. Yue, G.; Li, F.; Tan, F.; Li, G.; Chen, C.; Wu, J. Nickel sulfide films with significantly enhanced electrochemical performance induced by self-assembly of 4-aminothiophenol and their application in dye-sensitized solar cells. RSC Adv. 2014, 4, 64068-64074.