2-Nitrobenzamide, 98%
2-Nitrobenzamide, 98%
2-Nitrobenzamide, 98%
2-Nitrobenzamide, 98%
Thermo Scientific Chemicals

2-Nitrobenzamide, 98%

CAS: 610-15-1 | C7H6N2O3 | 166.136 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A14084.18
also known as A14084-18
Price (USD)
82.65
Online Exclusive
91.60
Save 8.95 (10%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
82.65
Online Exclusive
91.60
Save 8.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS610-15-1
IUPAC Name2-nitrobenzamide
Molecular FormulaC7H6N2O3
InChI KeyKLGQWSOYKYFBTR-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)170.0-176.0°C
Appearance (Color)White to pale cream
Water Content (Karl Fischer Titration)≤1%
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only