1-Bromonaphthalene, 97%
1-Bromonaphthalene, 97%
1-Bromonaphthalene, 97%
1-Bromonaphthalene, 97%
Thermo Scientific Chemicals

1-Bromonaphthalene, 97%

CAS: 90-11-9 | C10H7Br | 207.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A14122.0B
Price (USD)
277.65
Special offer
Online exclusive
Ends: 30-Jun-2026
326.00
Save 48.35 (15%)
Each
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)
277.65
Special offer
Online exclusive
Ends: 30-Jun-2026
326.00
Save 48.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS90-11-9
IUPAC Name1-bromonaphthalene
Molecular FormulaC10H7Br
InChI KeyDLKQHBOKULLWDQ-UHFFFAOYSA-N
SMILESBrC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to yellow to pale brown
Identification (FTIR)Conforms
Refractive Index1.6550-1.6600 @ 20?C
1-Bromonaphthalene is used in organic synthesis and refractometry of fats. It is used in the preparation of N-aryl imidazoles and diaryl ethers. It is used to determine the refractive index of crystals and the water content of alcohols. It is utilized in the preparation of glutathione peroxidase-like antioxidant activity of diaryl diselenides. Further, it serves as a solvent for the exfoliation and dispersion of hexabenzocoronene. In addition to this, it is used as a refrigerant and a solvent molecular weight substance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromonaphthalene is used in organic synthesis and refractometry of fats. It is used in the preparation of N-aryl imidazoles and diaryl ethers. It is used to determine the refractive index of crystals and the water content of alcohols. It is utilized in the preparation of glutathione peroxidase-like antioxidant activity of diaryl diselenides. Further, it serves as a solvent for the exfoliation and dispersion of hexabenzocoronene. In addition to this, it is used as a refrigerant and a solvent molecular weight substance.

Solubility
Miscible with chloroform, ethanol, ether, benzene and ethyl acetate. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Motogaito, A.; Nakamura, S.; Miyazaki, J.; Miyake, H.; Hiramatsu, K. Using surface-plasmon polariton at the GaP-Au interface in order to detect chemical species in high-refractive-index media. Opt. Commun. 2015, 341, 64-68.
  2. Gentry, S. P.; Halloran, J. W. Light scattering in absorbing ceramic suspensions: Effect on the width and depth of photopolymerized features. J. Eur. Ceram. Soc. 2015, 35 (6), 1895-1904.