(1R,2S)-(+)-10,2-Camphorsultam, 99%
(1R,2S)-(+)-10,2-Camphorsultam, 99%
(1R,2S)-(+)-10,2-Camphorsultam, 99%
Thermo Scientific Chemicals

(1R,2S)-(+)-10,2-Camphorsultam, 99%

CAS: 108448-77-7 | C10H17NO2S | 215.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A14133.06
also known as A14133-06
Price (USD)
232.65
Special offer
Online exclusive
Ends: 30-Jun-2026
274.00
Save 41.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
232.65
Special offer
Online exclusive
Ends: 30-Jun-2026
274.00
Save 41.35 (15%)
Each
Chemical Identifiers
CAS108448-77-7
IUPAC Name10,10-dimethyl-3λ⁶-thia-4-azatricyclo[5.2.1.0¹,⁵]decane-3,3-dione
Molecular FormulaC10H17NO2S
InChI KeyDPJYJNYYDJOJNO-UHFFFAOYNA-N
SMILESCC1(C)C2CCC11CS(=O)(=O)NC1C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Formcrystalline powder
Assay (GC)>98.5%
Optical Rotation+33 ? 1? (c=1, Ethanol)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only