1-Ethyl-4-iodobenzene, 98%
1-Ethyl-4-iodobenzene, 98%
1-Ethyl-4-iodobenzene, 98%
Thermo Scientific Chemicals

1-Ethyl-4-iodobenzene, 98%

CAS: 25309-64-2 | C8H9I | 232.064 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A14138.18
also known as A14138-18
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
113.00
Save 16.35 (14%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
113.00
Save 16.35 (14%)
Each
Chemical Identifiers
CAS25309-64-2
IUPAC Name1-ethyl-4-iodobenzene
Molecular FormulaC8H9I
InChI KeyOOLSRHZMXAYDFB-UHFFFAOYSA-N
SMILESCCC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
FormLiquid
Refractive Index1.5890-1.5930 @ 20?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only