2-Decanone, 97%
2-Decanone, 97%
2-Decanone, 97%
Thermo Scientific Chemicals

2-Decanone, 97%

CAS: 693-54-9 | C10H20O | 156.269 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14145.22
also known as A14145-22
Price (USD)
214.65
Special offer
Online exclusive
Ends: 30-Jun-2026
252.00
Save 37.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
214.65
Special offer
Online exclusive
Ends: 30-Jun-2026
252.00
Save 37.35 (15%)
Each
Chemical Identifiers
CAS693-54-9
IUPAC Namedecan-2-one
Molecular FormulaC10H20O
InChI KeyZAJNGDIORYACQU-UHFFFAOYSA-N
SMILESCCCCCCCCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
Refractive Index1.4230-1.4280 @ 20?C
Appearance (Color)Clear colorless
FormLiquid
2-Decanone was used in the synthesis of brushed block copolymers via conjugation through an acid-labile hydrazone linker to poly(ethylene glycol)-poly(aspartate hydrazide) block copolymers; methyl ketones. 2-decanone used as a solvent and an intermediate in the synthesis of pharmaceuticals and agrochemicals. It is also used in perfumes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Decanone was used in the synthesis of brushed block copolymers via conjugation through an acid-labile hydrazone linker to poly(ethylene glycol)-poly(aspartate hydrazide) block copolymers; methyl ketones. 2-decanone used as a solvent and an intermediate in the synthesis of pharmaceuticals and agrochemicals. It is also used in perfumes.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Dr. Eric Monflier; Dr. Emilie Blouet; Prof. Dr. Yolande Barbaux; and Prof. Dr. André Mortreux. Wacker Oxidation of 1-Decene to 2-Decanone in the Presence of a Chemically Modified Cyclodextrin System: A Happy Union of Host-Guest Chemistry and Homogeneous Catalysis. Angewandte Chemie International Edition in English. 1994, 33 (20), 2100-2102.
  2. Krysztof Januszkiewicz; Howard Alper. Palladium and phase transfer catalyzed oxidation of olefins to ketones. Sensitivity of the reaction to the nature of the phase transfer agent. Tetrahedron Letters. 1983, 24 (47), 5159-5162.