2-Methylpyrazine, 99+%
2-Methylpyrazine, 99+%
2-Methylpyrazine, 99+%
Thermo Scientific Chemicals

2-Methylpyrazine, 99+%

CAS: 109-08-0 | C5H6N2 | 94.117 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A14146.14
also known as A14146-14
Price (USD)
29.65
Online exclusive
32.60
Save 2.95
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
29.65
Online exclusive
32.60
Save 2.95
Each
Chemical Identifiers
CAS109-08-0
IUPAC Name2-methylpyrazine
Molecular FormulaC5H6N2
InChI KeyCAWHJQAVHZEVTJ-UHFFFAOYSA-N
SMILESCC1=CN=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥99.0%
Identification (FTIR)Conforms
Refractive Index1.5040-1.5060 @ 20?C
2-Methylpyrazine is used as flavoring agents in bread fresh bread, butter, chocolate, chocolate cocoa, coffee, meat roasted, nut almond.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylpyrazine is used as flavoring agents in bread fresh bread, butter, chocolate, chocolate cocoa, coffee, meat roasted, nut almond.

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
RUO – Research Use Only
Lucid Forni; Paolo Pollesel. TPD-TPR-MS mechanistic study of the synthesis of 2-methylpyrazine over palladized Zn-Cr oxide. Journal of Catalysis. 1991, 130 403-410.