Sebacic acid, 98+%
Sebacic acid, 98+%
Sebacic acid, 98+%
Thermo Scientific Chemicals

Sebacic acid, 98+%

CAS: 111-20-6 | C10H18O4 | 202.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1000 g
5000 g
Catalog number A14158.0B
also known as A14158-0B
Price (USD)
86.65
Special offer
Online exclusive
Ends: 28-Dec-2026
102.00
Save 15.35
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
86.65
Special offer
Online exclusive
Ends: 28-Dec-2026
102.00
Save 15.35
Each
Chemical Identifiers
CAS111-20-6
IUPAC Namedecanedioic acid
Molecular FormulaC10H18O4
InChI KeyCXMXRPHRNRROMY-UHFFFAOYSA-N
SMILESOC(=O)CCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder or granules or prills or beads
Assay (Silylated GC)≥98.0%
Sebacic acid is used as a monomer for nylon 610, plasticizers, lubricants, hydraulic fluids, cosmetics and candles. It is also used as intermediates in antiseptics, aromatics and painting materials. It is used in the synthesis of poly(glycerol sebacate) and novel bio-nylon. It finds application as adhesives, corrosion inhibitors and solvents. Its derivative, diethyl Sebacate is used in plastics manufacturing and engineering, as well as drug delivery procedures through the use of biodegradable encapsulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sebacic acid is used as a monomer for nylon 610, plasticizers, lubricants, hydraulic fluids, cosmetics and candles. It is also used as intermediates in antiseptics, aromatics and painting materials. It is used in the synthesis of poly(glycerol sebacate) and novel bio-nylon. It finds application as adhesives, corrosion inhibitors and solvents. Its derivative, diethyl Sebacate is used in plastics manufacturing and engineering, as well as drug delivery procedures through the use of biodegradable encapsulations.

Solubility
Slightly soluble in water.

Notes
Incompatible with bases, reducing agents and oxidizing agents.
RUO – Research Use Only

General References:

  1. Yabalak, E.; Gormez, O.; Gozmen, B.; Gizir, A. M. The solubility of sebacic acid in subcritical water using the response surface methodology. Int. J. Ind. Chem. 2015, 6 (1), 23-29.
  2. Kang, Y.; Chen, Z.; Wang, B.; Yang, Y. Synthesis and mechanical properties of thermoplastic films from lignin, sebacic acid and poly (ethylene glycol). Ind. Crops Prod. 2014, 56, 105-112.