3-Chloro-6-methoxypyridazine, 97%
3-Chloro-6-methoxypyridazine, 97%
3-Chloro-6-methoxypyridazine, 97%
3-Chloro-6-methoxypyridazine, 97%
Thermo Scientific Chemicals

3-Chloro-6-methoxypyridazine, 97%

CAS: 1722-10-7 | C5H5ClN2O | 144.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14173.06
also known as A14173-06
Price (USD)
56.65
Online Exclusive
63.20
Save 6.55 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
56.65
Online Exclusive
63.20
Save 6.55 (10%)
Each
Add to cart
Chemical Identifiers
CAS1722-10-7
IUPAC Name3-chloro-6-methoxypyridazine
Molecular FormulaC5H5ClN2O
InChI KeyXBJLKXOOHLLTPG-UHFFFAOYSA-N
SMILESCOC1=CC=C(Cl)N=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)88.5-94.5?C
Appearance (Color)White
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only