4-(Trifluoromethyl)benzoyl chloride, 97%
4-(Trifluoromethyl)benzoyl chloride, 97%
4-(Trifluoromethyl)benzoyl chloride, 97%
Thermo Scientific Chemicals

4-(Trifluoromethyl)benzoyl chloride, 97%

CAS: 329-15-7 | C8H4ClF3O | 208.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A14176.06
also known as A14176-06
Price (USD)
55.65
Special offer
Online exclusive
Ends: 28-Dec-2026
65.70
Save 10.05
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
55.65
Special offer
Online exclusive
Ends: 28-Dec-2026
65.70
Save 10.05
Each
Chemical Identifiers
CAS329-15-7
IUPAC Name4-(trifluoromethyl)benzoyl chloride
Molecular FormulaC8H4ClF3O
InChI KeyOXZYBOLWRXENKT-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(C=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4740-1.4790 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
4-(Trifluoromethyl)benzoyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzoyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store under dry inert gas. Protect from humidity and water. Incompatible with water, bases, alcohol and oxidizing agents.
RUO – Research Use Only

General References:

  1. Viera Poláčková; Štefan Toma; Iveta Augustínová. Microwave-promoted cross-coupling of acid chlorides with arylboronic acids: a convenient method for preparing aromatic ketones. Tetrahedron.2006, 62 11675-11678.